Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (1-Cyanoethyl)Phosphonate

Catalog No. 4085735G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-857-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 40-857-35G Supplier Sigma Aldrich Supplier No. 4085735G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reacant for:Preparation of sphingosine phosphate receptor (S1P) agonists with N-cinnamyl-+â-alanine moietyPreparation of benzyl- and benzoyldimethylaminopyridinones as HIV-1 reverse transcriptase inhibitorsStereoselective preparation of cyano(trifluoromethyl)pentenenitriles via alkylation, followed by elimination reactionSynthesis of [(methylenepiperidinyl) or (methylenepyrrolidinyl)fluorophenyl]oxazolidinone antibacterial agentsHorner-Wadsworth-Emmons olefination

Specifications

CAS 10534-59-5
Density 0.99 g/mL at 25°C
Boiling Point 100°C
Molecular Formula C18H39NO2
Refractive Index n20/D 1.432
Linear Formula [CH3(CH2)3]4N(O2CCH3)
MDL Number MFCD00043208
Quantity 5 g
Synonym (1-Cyanoethyl)phosphonic acid diethyl ester; NSC 135193
Molecular Weight (g/mol) 301.51 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.