Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Azodicarboxylate Solution

Catalog No. 56311025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-311-025G 25 g
56-311-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 56-311-025G Supplier Sigma Aldrich Supplier No. 56311025G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DOT Special Approval Submission is Under Review. Availability is Estimated as March 2012.Reactant for preparation of:Immunostimulants +a-GalactosylceramidesCellotriose and cellotetraose analogues as transition state mimics for mechanistic studies of cellulasesBisubstrate inhibitors with molecular recognition at the active site of catechol-O-methyltransferaseDerivatives of F200 and S383 with cannabinoid CB1 receptor binding activitiesAza-+a-lactams via NHC-catalyzed [2 + 2] cycloaddition with ketenesReagent for:Annulation of N-protected imines+a-thiocyanation of enolizable ketones with ammonium thiocyanateDiels-Alder reactions

Specifications

CAS 1972-28-7
Density 0.956 g/mL at 25°C
Boiling Point 116°C to 117°C
Molecular Formula C6H10N2O4
Linear Formula C2H5OCON=NCOOC2H5
Refractive Index n20/D 1.4690
MDL Number MFCD00009103
Quantity 25 g
Synonym 1,2-Ethoxycarbonyl diazene solution; DEAD; Diethoxycarbonyldiazene solution; Diethyl azodiformate solution; NSC 3474; NSC 679015; Unifoam AZ-AE 200
Molecular Weight (g/mol) 174.15 g/mol
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.