Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D377525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D377525G 25 g
D37751G 1 g
D37755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D377525G Supplier TCI America Supplier No. D377525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1149-23-1
Molecular Formula C13H19NO4
Molecular Weight (g/mol) 253.298
MDL Number MFCD00005951
InChI Key LJXTYJXBORAIHX-UHFFFAOYSA-N
Synonym diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
PubChem CID 70849
IUPAC Name diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C

Specifications

Melting Point 183°C
Quantity 25 g
Formula Weight 253.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
RTECSNumber US7970100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.