Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2-(2-cyanoethyl)malonate, 98%, Thermo Scientific™

Catalog No. AC170420050
Click to view available options
:
5g; Glass bottle

Specifications

CAS 17216-62-5
Density 1.0700g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C10H15NO4
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Linear Formula NCCH2CH2CH(CO2C2H5)2
Quantity 5g
MDL Number MFCD00001966
Synonym diethyl 2-2-cyanoethyl malonate, diethyl 2-cyanoethyl malonate, 1,3-diethyl 2-2-cyanoethyl propanedioate, diethyl2-2-cyanoethyl malonate, diethyl 2-2-cyanoethyl-malonate, propanedioic acid, 2-cyanoethyl-, diethyl ester, 2-cyanoethylmalonic acid diethylester, diethyl 2-2-cyanoethyl propane-1,3-dioate, pubchem14162
InChI Key YJJLOESDBPRZIP-UHFFFAOYSA-N
SMILES CCOC(=O)C(CCC#N)C(=O)OCC
IUPAC Name diethyl 2-(2-cyanoethyl)propanedioate
Molecular Weight (g/mol) 213.233
PubChem CID 87005
Formula Weight 213.23
Percent Purity ≥97.5%
Chemical Name or Material Diethyl 2-(2-cyanoethyl)malonate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-260-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.