Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 97%, Thermo Scientific™

Catalog No. AC114280250
Click to view available options
:
25g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2436-79-5
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H17NO4
Quantity 25g
MDL Number MFCD00005218
Synonym diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate
InChI Key XSBSXJAYEPDGSF-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C
IUPAC Name diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
Molecular Weight (g/mol) 239.271
PubChem CID 75526
Formula Weight 239.27
Percent Purity ≥96.0%
Chemical Name or Material Diethyl 2, 4-dimethylpyrrole-3, 5-dicarboxylate, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-431-2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.