Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (2-Oxo-2-phenylethyl)phosphonate 97.0+%, TCI America™
SDP

Catalog No. D33391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33391G 1 g
D33395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33391G Supplier TCI America Supplier No. D33391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3453-00-7
Molecular Formula C12H17O4P
Molecular Weight (g/mol) 256.24
MDL Number MFCD00015342
InChI Key HPEVTTNSIPGLEL-UHFFFAOYSA-N
Synonym Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester
PubChem CID 372803
IUPAC Name diethyl (2-oxo-2-phenylethyl)phosphonate
SMILES CCOP(=O)(CC(=O)C1=CC=CC=C1)OCC

Specifications

Color Yellow
Boiling Point 193°C
Quantity 1 g
Formula Weight 256.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (2-Oxo-2-phenylethyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.