Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 3-oxopimelate, 97%, Thermo Scientific™

Catalog No. AC213950050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40420-22-2
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00009214
InChI Key KAMXUPAAAGYEDK-UHFFFAOYSA-N
Synonym diethyl 3-oxopimelate, diethyl3-oxopimelate, heptanedioic acid, 3-oxo-, diethyl ester, 1,7-diethyl 3-oxoheptanedioate, 3-oxopimelic acid diethyl ester, diethyl3-oxoheptanedioate, acmc-1akc9, diethyl-3-oxopimelate, diethyl 3-oxoheptane-1,7-dioate
PubChem CID 575510
IUPAC Name 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate
SMILES CCOC(=O)CC(=O)C(C)(C)C(=O)OCC

Specifications

Color Brown-Yellow to Yellow
Density 1.08g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4427 to 1.4447 (20°C, 589nm)
Linear Formula C2H5O2C(CH2)3COCH2CO2C2H5
Quantity 5 g
Beilstein 03, III, 1380
Formula Weight 230.26
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Diethyl 3-oxopimelate, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.