Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 3-Oxopimelate 96.0+%, TCI America™
SDP

Catalog No. O02291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02291G 1 g
O02295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02291G Supplier TCI America Supplier No. O02291G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40420-22-2
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00009214
InChI Key KAMXUPAAAGYEDK-UHFFFAOYSA-N
Synonym diethyl 3-oxopimelate, diethyl3-oxopimelate, heptanedioic acid, 3-oxo-, diethyl ester, 1,7-diethyl 3-oxoheptanedioate, 3-oxopimelic acid diethyl ester, diethyl3-oxoheptanedioate, acmc-1akc9, diethyl-3-oxopimelate, diethyl 3-oxoheptane-1,7-dioate
PubChem CID 575510
IUPAC Name 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate
SMILES CCOC(=O)CC(=O)C(C)(C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 132°C
Quantity 1 g
Formula Weight 230.26
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Diethyl 3-Oxopimelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.