Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-Methylbenzyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. D333625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D333625G 25 g
D33365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D333625G Supplier TCI America Supplier No. D333625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3762-25-2
Molecular Formula C12H19O3P
Molecular Weight (g/mol) 242.255
MDL Number MFCD00041385
InChI Key QKGBKPZAXXBLJE-UHFFFAOYSA-N
Synonym diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
PubChem CID 3050
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methylbenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C)OCC

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 242.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Methylbenzyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.