Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 5-Amino-3-methyl-2,4-thiophenedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. A135425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A135425G 25 g
A13545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A135425G Supplier TCI America Supplier No. A135425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4815-30-9
Molecular Formula C11H15NO4S
Molecular Weight (g/mol) 257.30
MDL Number MFCD00005450
InChI Key DGVXLHAJVRRLGV-UHFFFAOYSA-N
Synonym diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate, diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate, diethyl 5-amino-3-methyl-thiophene-2,4-dicarboxylate, diethyl5-amino-3-methylthiophene-2,4-dicarboxylate, 2,4-thiophenedicarboxylic acid, 5-amino-3-methyl-, diethyl ester, 2-amino-3,5-bis ethoxycarbonyl-4-methylthiophene, 5-amino-3-methyl-thiophene-2,4-dicarboxylic acid diethyl ester, 2-amino-4-methyl-3,5-bis ethoxycarbonyl thiophene, diethyl 5-amino-3-methyl-2,4-thiophene dicarboxylate
PubChem CID 78537
IUPAC Name 2,4-diethyl 5-amino-3-methylthiophene-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(C)C(C(=O)OCC)=C(N)S1

Specifications

Melting Point 108°C
Quantity 25 g
Formula Weight 257.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 5-Amino-3-methyl-2,4-thiophenedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.