Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Acetamidomalonate, 99+%

Catalog No. p-108168
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

CAS: 1068-90-2 | C9H15NO5 | 217.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-90-2
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.22
MDL Number MFCD00009146
InChI Key ISOLMABRZPQKOV-UHFFFAOYSA-N
Synonym diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
PubChem CID 14041
IUPAC Name diethyl 2-acetamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC(=O)C

Specifications

Melting Point 94°C to 97°C
Color White to Yellow
Boiling Point 185°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C2H5O2CCH(NHCOCH3)CO2C2H5
Quantity 25 g
Beilstein 04, II, 891
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 217.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Diethyl acetamidomalonate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 213-952-9
RTECSNumber OO0360000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.