Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl acetylsuccinate, 97+%

Catalog No. AAB2374414
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1115-30-6 | C10H16O5 | 216.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC

Specifications

Melting Point -8°C
Density 1.081
Boiling Point 180°C to 183°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4346
Quantity 25 g
Formula Weight 216.23
Percent Purity ≥97%
Chemical Name or Material Diethyl acetylsuccinate
EINECSNumber 214-223-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.