Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl chlorophosphate, 97+%

Catalog No. aah3382014
Click to view available options
Quantity:
25 g
100 g

CAS: 814-49-3 | C4H10ClO3P | 172.545 g/mol

A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-49-3
Molecular Formula C4H10ClO3P
Molecular Weight (g/mol) 172.545
MDL Number MFCD00009075
InChI Key LGTLXDJOAJDFLR-UHFFFAOYSA-N
Synonym diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
PubChem CID 13139
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)Cl

Specifications

Density 1.194
Boiling Point 60°C (2 mmHg)
Flash Point 61°C (142°F)
Quantity 25 g
UN Number UN3278
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 172.55
Percent Purity ≥97%
Chemical Name or Material Diethyl chlorophosphate
Hazard Category H300+H310+H330-H314
Hazard Statement GHS H Statement
H300-H310-H330-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
EINECSNumber 212-369-4
RTECSNumber TD1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.