Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyl cyanophosphonate, 93%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)

Specifications
Specifications
Color | Colorless to Yellow |
Density | 1.0700g/mL |
Boiling Point | 104.0°C to 105.0°C (19.0 mmHg) |
Flash Point | 80°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 91% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4020 to 1.4040 |
Linear Formula | (C2H5O)2P(O)CN |
Quantity | 25 g |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Fatal in contact with skin. Fatal if swallowed. Fatal if inhaled. Contact with acids liberates very toxic gas. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Gently wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 220-936-5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction