Learn More
Diethyl diallylmalonate, 97+%
Description
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke?s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.
Solubility
Sparingly Soluble in water. (0.72 g/L) (25°C)
Notes
Store under cool dry place. Ensure good ventilation.

Specifications
Specifications
Density | 0.993 |
Boiling Point | 242°C to 244°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.446 |
Quantity | 50 g |
Beilstein | 1790529 |
Synonym | diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69 |
Solubility Information | Sparingly Soluble in water. (0.72g/L) (25°C) |
Formula Weight | 240.3 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 221-696-4 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.