Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl diethylmalonate, 98%

Catalog No. AAB2381022
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 77-25-8 | C11H20O4 | 216.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-25-8
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00009132
InChI Key ZKBBUZRGPULIRN-UHFFFAOYSA-N
Synonym diethyl diethylmalonate, diethyl 2,2-diethylmalonate, diethylmalonic ester, propanedioic acid, diethyl-, diethyl ester, diethylmalonate diethyl ester, diethylmalonic acid diethyl ester, diethylmalonic acid, diethyl ester, unii-ceh13944yq, 1,3-diethyl 2,2-diethylpropanedioate, diethyl diethylmalonate diethyl
PubChem CID 66165
IUPAC Name diethyl 2,2-diethylpropanedioate
SMILES CCC(CC)(C(=O)OCC)C(=O)OCC

Specifications

Density 0.99
Boiling Point 228°C to 230°C
Flash Point 94°C (202°F)
Refractive Index 1.423
Quantity 100 g
Merck Index 14,3789
Formula Weight 216.28
Percent Purity 98%
Chemical Name or Material Diethyl diethylmalonate
EINECSNumber 201-016-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.