Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl dimethylaminomethylenemalonate, 94%

Catalog No. AAB2076506
Click to view available options
Quantity:
5 g

CAS: 18856-68-3 | C10H17NO4 | 215.25 g/mol

Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18856-68-3
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD00052394
InChI Key XNSXXTKUKYWLAH-UHFFFAOYSA-N
Synonym diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
PubChem CID 140439
IUPAC Name diethyl 2-(dimethylaminomethylidene)propanedioate
SMILES CCOC(=O)C(=CN(C)C)C(=O)OCC

Specifications

Boiling Point 134°C to 136°C (0.5 mmHg)
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material Diethyl dimethylaminomethylenemalonate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.