Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, 99+%, for spectroscopy, stabilized with ethanol

Catalog No. p-3598357
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle

CAS: 60-29-7 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.0°C
Absorbance 0.004 max. at 400nm, 0.005 max. at 280nm, 0.03 max. at 260nm, 0.1 max. at 245nm, 0.4 max. at 230nm, 1 max. at 220nm
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 500 g
Flash Point −45°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3510 to 1.3530
Assay Percent Range 99+%
Specific Gravity 0.714
Linear Formula (CH3CH2)2O
Viscosity 0.2448 mPa.s (20°C)
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Formula Weight 74.12
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Diethyl ether, Stabilized with ethanol, for spectroscopy
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.