Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%, Solstice

Catalog No. 60046381 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-381 1 L Glass Bottle
60-046-380 100 mL Glass Bottle
60-046-382 250 mL Glass Bottle
60-046-383 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-381 Supplier Solstice Supplier No. 3461366X1L

ACS Reagent, contains BHT as inhibitor, anhydrous,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form anhydrous
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99%
Grade ACS
Assay (GC)
Chemical Name or Material Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
EINECSNumber 200-467-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.