Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Ether, puriss., ≥99.5% (GC), Solstice

Catalog No. 60045502 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
135 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-502 135 kg Metal Drum
60-045-503 1 L Glass Bottle
60-045-510 5 L Aluminum Bottle
60-045-512 6 x 1 L Glass Bottle
60-045-508 4 x 2.5 L Glass Bottle
60-045-509 4 x 5 L Aluminum Bottle
60-045-504 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-045-502 Supplier Solstice Supplier No. 24004135KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Density 0.706g/mL at 25°C
Quantity 135 kg
Explosion Limit 0.365
Flash Point −45°C (−49°F)
Ignition Point 320°F
Packaging Metal Drum
Physical Form Liquid
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34.6°C
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Diethyl Ether, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Hazard Statement P210-P261
Precautionary Statement H224-H302-H336
DOTInformation class 3 - PG 1 - Diethyl ether
EINECSNumber 200-467-2
RTECSNumber KI5775000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.