Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Ether, technical, stabilized

Catalog No. ac444260050
Click to view available options
Quantity:
5 L
Packaging:
Metal Drum

CAS: 60-29-7 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.0°C
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 5 L
Flash Point −45°C
Packaging Metal Drum
Refractive Index 1.3510 to 1.3530
Specific Gravity 0.714
Linear Formula (CH3CH2)2O
Viscosity 0.2448 mPa.s (20°C)
Merck Index 15,3861
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid,miscible with lower aliphatic alcohols,benzene,,chloroform,petroleum ether and many oils
Formula Weight 74.12
Grade Technical
Chemical Name or Material Diethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No
EINECSNumber 200-467-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.