Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl hexafluoroglutarate, 97%, Thermo Scientific Chemicals

Catalog No. AAB2242806
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 424-40-8
Molecular Formula C9H10F6O4
Molecular Weight (g/mol) 296.17
MDL Number MFCD00015156
InChI Key MSDPXVBLFJODJO-UHFFFAOYSA-N
Synonym diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester
PubChem CID 9874
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC

Specifications

Density 1.365
Boiling Point 224°C
Refractive Index 1.36
Quantity 5 g
Beilstein 1804802
IUPAC Name diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Formula Weight 296.16
Percent Purity 97%
Chemical Name or Material Diethyl hexafluoroglutarate
Hazard Category H227-H315-H319-H332-H335
Hazard Statement GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-035-2
RTECSNumber MA3750500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.