Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyl (Hydroxymethyl)phosphonate 97.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 105°C |
Quantity | 100 g |
Formula Weight | 168.13 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Diethyl (Hydroxymethyl)phosphonate |
Safety and Handling
TSCA | Yes |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction