Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl iodomethylphosphonate, 98+%

Catalog No. AAA1369714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1369714 25 g
AAA1369706 5 g
AAA1369722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1369714 Supplier Thermo Scientific Chemicals Supplier No. A1369714
Add to Cart

CAS: 10419-77-9 | C5H12IO3P | 278.03 g/mol

Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10419-77-9
Molecular Formula C5H12IO3P
Molecular Weight (g/mol) 278.03
MDL Number MFCD00041386
InChI Key PNFHTBDRPOJJTQ-UHFFFAOYSA-N
Synonym diethyl iodomethylphosphonate, diethyl iodomethyl phosphonate, phosphonic acid, iodomethyl-, diethyl ester, acmc-20al8r, diethyliodomethylphosphonate, diethyl iodomethanephosphonate, iodomethylphosphonic acid diethyl ester, diethyl iodomethylphosphonate 5g, iodomethyl-phosphonic acid diethyl ester
PubChem CID 82616
IUPAC Name 1-[ethoxy(iodomethyl)phosphoryl]oxyethane
SMILES CCOP(=O)(CI)OCC

Specifications

Boiling Point 112°C to 114°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4995
Quantity 25 g
Beilstein 1812052
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 278.03
Percent Purity ≥98%
Chemical Name or Material Diethyl iodomethylphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-895-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.