Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl isopropylidenemalonate, 97%

Catalog No. AC443310050
Click to view available options
Quantity:
5 g

CAS: 6802-75-1 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6802-75-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
InChI Key WEISAZNMMVPNTH-UHFFFAOYSA-N
Synonym diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m
PubChem CID 81255
IUPAC Name diethyl 2-propan-2-ylidenepropanedioate
SMILES CCOC(=O)C(=C(C)C)C(=O)OCC

Specifications

Density 1.0210g/mL
Boiling Point 175°C to 178°C (120.0 mmHg)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.021
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Diethyl isopropylidenemalonate
EINECSNumber 229-876-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.