Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl L-Glutamate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. G017925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G017925G 25 g
G01795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G017925G Supplier TCI America Supplier No. G017925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00012509
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 114°C
Color White
Quantity 25 g
Formula Weight 239.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl L-Glutamate Hydrochloride
EINECSNumber (2)-2712
RTECSNumber MA1252280
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.