Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxylamine hydrochloride, 97%
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Melting Point | 155°C to 158°C |
Color | White |
Physical Form | Crystalline Powder, Crystals and/or Chunks |
Quantity | 1 kg |
Assay Percent Range | 97% |
Linear Formula | H2NOH·HCl |
Fieser | 01,478; 02,217; 07,176; 09,245 |
Merck Index | 15,4867 |
Solubility Information | Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether |
Formula Weight | 69.49 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May be corrosive to metals. Toxic if swallowed. Harmful in contact with skin. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Suspected of causing canc |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Wear protecti |
EINECSNumber | 226-798-2 |
RTECSNumber | NC3675000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction