Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl malonate, 99%

Manufacturer:  Alfa Aesar A1546836

Quantity 500g

 View more versions of this product

Catalog No. AAA1546836


  Check Availability
Add to cart

Specifications

Description

Applications
Diethyl malonate is used in organic synthesis for the preparation of alpha-aryl malonates, mono-substituted and di-substituted acetic acid, barbiturates and artificial flavorings. It is also involved in the synthesis of pharmaceuticals like chloroquine, butazolidin and barbital. It acts as intermediate in the synthesis of vitamin B1, vitamin B6, non-steroidal anti-inflammatory agents agrochemicals and perfumes. In Knoevenagel condensation reaction, it reacts with benzaldehyde to get diethyl benzylidenemalonate.

Solubility
Miscible with ethyl alcohol, ether, chloroform and benzene. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Description & Specifications

Chemical Identifiers

105-53-3
160.17
774687
Diethyl Malonate
CH2(CO2CH2CH3)2
MFCD00009195
14,3823

Specifications

500g
1.055
99%
160.17
774687
Diethyl Malonate
105-53-3
Liquid
CH2(CO2CH2CH3)2
MFCD00009195
14,3823
99%

Safety and Handling

SDS & Safety Documents

Warning

RTECSNumber : OO0700000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit