Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (Methoxymethyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. D387325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. D387325G

Supplier: TCI America D387325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32806-04-5
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00137944
InChI Key IWEOGHGFCXMKFM-UHFFFAOYSA-N
Synonym (Methoxymethyl)phosphonic Acid Diethyl Ester
PubChem CID 11658394
IUPAC Name diethyl (methoxymethyl)phosphonate
SMILES CCOP(=O)(COC)OCC

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 g
Formula Weight 182.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Methoxymethyl)phosphonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.