Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (Methylthiomethyl)phosphonate 97.0+%, TCI America™
SDP

Catalog No. M120825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M120825G 25 g
M12085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M120825G Supplier TCI America Supplier No. M120825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28460-01-7
Molecular Formula C6H15O3PS
Molecular Weight (g/mol) 198.217
MDL Number MFCD00015131
InChI Key RAABCCZMKIIGJW-UHFFFAOYSA-N
Synonym diethyl methylthiomethylphosphonate, diethyl methylthiomethyl phosphonate, diethyl methylthio methyl phosphonate, diethyl methylsulfanyl methylphosphonate, methylthiomethyl phosphonic acid diethyl ester, 1-ethoxy methylsulfanylmethyl phosphoryl oxyethane, phosphonic acid, methylthio methyl-, diethyl ester, diethoxy methylthiomethyl phosphino-1-one, pubchem12686
PubChem CID 520377
IUPAC Name 1-[ethoxy(methylsulfanylmethyl)phosphoryl]oxyethane
SMILES CCOP(=O)(CSC)OCC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Methylthiomethyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.