Alfa Aesar™ Diethyl phosphite, 97+%

Manufacturer: Alfa Aesar™ 4400036

Quantity 500g

 View more versions of this product

Catalog No. AA4400036

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 762-04-9
    Molecular Formula (C2H5O)2P(O)H
    Formula Weight 138.1
    MDL Number MFCD00044573
    Chemical Name or Material Diethyl phosphite

    Specifications

    Formula Weight 138.1
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    Molecular Formula (C2H5O)2P(O)H
    CAS 762-04-9
    Flash Point 74°C (165°F)
    Sensitivity Moisture Sensitive
    Health Hazard 2 GHS H Statement
    H318-H317-H227
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Combustible liquid.
    RTECS Number TG7875000
    Density 1.070g/mL
    Boiling Point 50° to 51°C (2mmHg)
    Color Colorless
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under Argon
    Melting Point −70°C
    Quantity 500g
    MDL Number MFCD00044573
    Odor Weak
    Chemical Name or Material Diethyl phosphite
    Assay ≥97%

    GHS H Statement
    H318-H317-H227
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit