Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phosphite, 97+%, Thermo Scientific Chemicals

Catalog No. AA4400022
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg

Diethyl phosphite is precursor used for the synthesis of various organophosphorus compounds. used to prepare the nickel chloride-diethyl phosphite system, efficient catalyst for the cross-coupling reaction between various functionalized arylzinc halides and aryl bromides, triflates and activated chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-04-9
Molecular Formula C4H10O3P+
Molecular Weight (g/mol) 137.095
MDL Number MFCD00044573
InChI Key LXCYSACZTOKNNS-UHFFFAOYSA-N
Synonym diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
PubChem CID 6327654
IUPAC Name diethoxy(oxo)phosphanium
SMILES CCO[P+](=O)OCC

Specifications

Melting Point -70°C
Color Colorless
Density 1.070 g/mL
Boiling Point 50°C to 51°C (2 mmHg)
Flash Point 74°C (165°F)
Odor Weak
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 138.1
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Diethyl phosphite
Show More Show Less
Hazard Category H227-H317-H318
Hazard Statement GHS H Statement
H318-H317-H227
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
EINECSNumber 212-091-6
RTECSNumber TG7875000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.