Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Phosphoramidate 98.0+%, TCI America™
SDP

Catalog No. D236125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D236125G 25 g
D23615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D236125G Supplier TCI America Supplier No. D236125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1068-21-9
Molecular Formula C4H12NO3P
Molecular Weight (g/mol) 153.118
MDL Number MFCD00015676
InChI Key MCQILDHFZKTBOD-UHFFFAOYSA-N
Synonym Diethyl Amidophosphate, Diethyl Phosphoryl Amide
PubChem CID 70605
IUPAC Name [amino(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(N)OCC

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 153.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Phosphoramidate
EINECSNumber (2)-1992
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.