Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phthalate, 99%

Catalog No. AAA175290B
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA175290B 1000 g
AAA1752930 250 g
AAA175290E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA175290B Supplier Thermo Scientific Chemicals Supplier No. A175290B
Add to Cart

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol

Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Melting Point -64°C
Density 1.118
Boiling Point 296°C to 298°C
Flash Point 160°C (320°F)
Refractive Index 1.502
Quantity 1000 g
Beilstein 1912500
Merck Index 14,7371
Solubility Information Miscible with ethanol,chloroform,diethyl ether,benzene,acetone,vegetable oils,ketones,carbon tetrachloride,esters,aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Diethyl phthalate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-550-6
RTECSNumber TI1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.