Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Phthalate, Spectrum™ Chemical
SDP

Catalog No. 18620521 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-521 4 L
18-620-520 500 mL
18-620-522 20 L
18-620-523 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-521 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D10284LTGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84-66-2

Specifications

Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 99%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.