Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Propylmalonate 98.0+%, TCI America™
SDP

Catalog No. D470825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D470825ML 25 mL
D47085ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D470825ML Supplier TCI America Supplier No. D470825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2163-48-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009168
InChI Key GRRSDGHTSMJICM-UHFFFAOYSA-N
Synonym diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate
PubChem CID 16552
IUPAC Name diethyl 2-propylpropanedioate
SMILES CCCC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 222°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Propylmalonate
EINECSNumber (2)-0861
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.