Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyl Pyrocarbonate, >97%, Ultrapure
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
Description
- A strong, nonspecific inhibitor of ribonuclease
- Reacts with the N1 of histidine residues in proteins under mild conditions
- Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
- Reacts with amines and thus cannot be used with Tris Buffer
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Strong, nonspecific inhibitor of ribonuclease
- Reacts with N1 of histidine residues in proteins under mild conditions
- Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
- Reacts

Specifications
Specifications
Melting Point | 69°C |
Density | 1.101 |
Boiling Point | 93°C to 94°C (18 mmHg) |
Flash Point | 69°C |
Packaging | Plastic bottle |
Refractive Index | 1.398 |
Quantity | 100 mL |
Beilstein | 637031 |
Formula Weight | 162.14 |
Percent Purity | ≥97% |
Show More |
Safety and Handling
Hazard Category | H227-H302-H315-H319-H332-H335 |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 216-542-8 |
RTECSNumber | LQ9350000 |
TSCA | Yes |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction