Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Pyrocarbonate, MP Biomedicals™

Catalog No. MP215090280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215090280 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215090280 Supplier MP Biomedicals, Inc Supplier No. 0215090280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to inactivate RNase enzymes. Diethyl Pyrocarbonate, MP Biomedicals is sensitive to moisture and pH. DEPC is also sensitive to ammonia, which causes decomposition to urethane, a possible carcinogen. It protects RNA against degradation and is effective

DEPC inactivates RNase in solution at about 0.1% (v/v). It inhibits the ryanodine binding to ryanodine/Ca2+ receptor channels in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles. DEPC is effective as a nuclease inhibitor with a variety of uses:

  • Modification reagent for His and Tyr residues in proteins
  • Robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases
  • Used to modify proteins and nucleic acids
  • PCR reaction for treating deionized water, which reduces the risk of RNA being degraded by RNases
  • Dot blot hybridization, to dilute total RNA isolated from different micro-organisms
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name ethoxycarbonyl ethyl carbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Density 1.101g/mL at 25°C (Lit.)
Boiling Point 93°C to 94°C (18 mmHg)(literature)
Refractive Index n20/D 1.398 (Lit.)
Quantity 100 mL
Solubility Information Soluble in alcohols, esters, ketones, hydrocarbons. A 50% w/w solution in ethanol may be prepared. Slowly soluble in water with hydrolytic decomposition yielding ethanol and CO2.
Formula Weight 162.10
Concentration or Composition (by Analyte or Components) Working Concentration: For Treatment of water to inhibit Rnase: Add 1mL DEPC to 1L water. Let sit overnight. Autoclave.
Physical Form Liquid
Recommended Storage Store at 4°C, under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.