Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Sulfite 98.0+%, TCI America™
SDP

Catalog No. D052825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D052825G 25 g
D0528500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D052825G Supplier TCI America Supplier No. D052825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-81-4
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.18
MDL Number MFCD00009098
InChI Key NVJBFARDFTXOTO-UHFFFAOYSA-N
Synonym sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester
PubChem CID 12197
IUPAC Name diethyl sulfite
SMILES CCOS(=O)OCC

Specifications

Color Colorless
Boiling Point 157°C
Quantity 25 g
UN Number 1993
Formula Weight 138.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Sulfite
RTECSNumber WT3511000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.