Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl terephthalate, 98%

Catalog No. p-109911
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 636-09-9 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 42.0°C to 45.0°C
Color White
Density 1.1100g/mL
Boiling Point 302.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ether,ethanol
Formula Weight 222.24
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Diethyl terephthalate, 98%
Show More Show Less
EINECSNumber 211-249-1
RTECSNumber WZ1223400
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.