Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate 96.0+%, TCI America™
SDP

Catalog No. D46111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46111G 1 g
D4611200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46111G Supplier TCI America Supplier No. D46111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71885-51-3
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD09263515
InChI Key RUPVGTAIRUGQIQ-UHFFFAOYSA-N
Synonym [(Tetrahydropyran-2-yloxy)methyl]phosphonic Acid Diethyl Ester
PubChem CID 11230495
IUPAC Name 2-(diethoxyphosphorylmethoxy)oxane
SMILES CCOP(=O)(COC1CCCCO1)OCC

Specifications

Color Yellow
Boiling Point 95°C
Quantity 1 g
Formula Weight 252.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.