Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18602195
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can

DI117, 109-89-7, C4H11N

Spectrum™ Chemical Diethylamine, Reagent is a secondary amine that is a flammable, volatile, strongly alkaline, colorless liquid that has a strong unpleasant smell. It is used in the production of rubber, resin, dyes and pharmaceuticals and as a corrosion inhibiter. Its Reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
IUPAC Name diethylamine
SMILES CCNCC

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 4 L
UN Number UN1154
Percent Purity 98%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.