Learn More
Diethylaminosulfur trifluoride, 95%
Description
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.
Solubility
Miscible with water, ethanol, benzene and acetonitrile.
Notes
Store in cool place. Moisture sensitive. It reacts with water and ethanol. Incompatible with water, acids and bases.

Specifications
Specifications
Density | 1.23 |
Boiling Point | 30°C to 32°C (3 mmHg) |
Flash Point | 23°C (73°F) |
Refractive Index | 1.4115 |
Linear Formula | (CH3CH2)2NSF3 |
Quantity | 25 g |
UN Number | UN2920 |
Beilstein | 1849066 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with water,ethanol,benzene and acetonitrile. |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H314-H335 |
Hazard Statement | GHS H Statement H331-H314-H318-H226 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
EINECSNumber | 253-771-2 |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.