Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethylaminosulfur trifluoride, 95%
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Color | Brown to Yellow |
| Density | 1.3160g/mL |
| Boiling Point | 30.0°C to 32.0°C (3.0 mmHg) |
| Flash Point | 23°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% |
| Packaging | Plastic bottle |
| Refractive Index | 1.4100 to 1.4160 |
| Linear Formula | (C2H5)2NSF3 |
| Quantity | 5 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Reacts violently with water. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open f |
| EINECSNumber | 253-771-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction