Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylaminosulfur trifluoride, 95%

Catalog No. p-4112307
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle

CAS: 38078-09-0 | C4H10F3NS | 161.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38078-09-0
Molecular Formula C4H10F3NS
Molecular Weight (g/mol) 161.18
MDL Number MFCD00000363
InChI Key CSJLBAMHHLJAAS-UHFFFAOYSA-N
Synonym diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
PubChem CID 123472
IUPAC Name N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
SMILES CCN(CC)S(F)(F)F

Specifications

Color Brown to Yellow
Density 1.3160g/mL
Boiling Point 30.0°C to 32.0°C (3.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Refractive Index 1.4100 to 1.4160
Linear Formula (C2H5)2NSF3
Quantity 5 g
Fieser 06,183; 10,142; 12,183; 13,110; 16,128
Specific Gravity 1.316
Merck Index 15, 3124
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,thf,ether,diglyme and,dichloroethylene
Formula Weight 161.18
Percent Purity 95%
Physical Form Fuming Liquid
Chemical Name or Material Diethylaminosulfur trifluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapour.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
EINECSNumber 253-771-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.