Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylbenzene (mixture) 98.0+%, TCI America™
SDP

Catalog No. p-7127702
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 25340-17-4
Molecular Formula C10H14
MDL Number MFCD00792900

Specifications

Melting Point -31°C
Color Colorless
Boiling Point 184°C
Quantity 25 mL
UN Number 2049
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylbenzene (mixture)
EINECSNumber (3)-0013&(3)-0060
RTECSNumber CZ5600000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.