Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol 2-Bromoethyl Methyl Ether 94.0+%, TCI America™
SDP

Catalog No. D38315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38315G 5 g
D383125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38315G Supplier TCI America Supplier No. D38315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72593-77-2
Molecular Formula C7H15BrO3
Molecular Weight (g/mol) 227.098
InChI Key LCCCTXULXHJDLA-UHFFFAOYSA-N
Synonym 2-[2-(2-Methoxyethoxy)ethoxy]ethyl Bromide, 1-Bromo-3,6,9-trioxadecane, mPEG3-Bromide
PubChem CID 13230355
IUPAC Name 1-[2-(2-bromoethoxy)ethoxy]-2-methoxyethane
SMILES COCCOCCOCCBr

Specifications

Boiling Point 65°C
Quantity 5 g
Formula Weight 227.10
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol 2-Bromoethyl Methyl Ether
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.