Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™
SDP

Catalog No. D1571100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1571100ML 100 mL
D157125ML 25 mL
D1571500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1571100ML Supplier TCI America Supplier No. D1571100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4246-51-9
Molecular Formula C10H26N2O3
Molecular Weight (g/mol) 222.33
MDL Number MFCD00059850
InChI Key JCEZOHLWDIONSP-UHFFFAOYSA-P
Synonym 4,7,10-trioxa-1,13-tridecanediamine, 3,3'-oxybis ethane-2,1-diyl bis oxy bis propan-1-amine, diethylene glycol bis 3-aminopropyl ether, unii-95w232h4rt, 3,3'-oxybis ethyleneoxy bis propylamine, di 3-aminopropyl ether of diethylene glycol, diethylene glycol, di 3-aminopropyl ether, 1-propanamine, 3,3'-oxybis 2,1-ethanediyloxy bis, 3,3'-oxybis 2,1-ethanediyloxy bis-1-propanamine
PubChem CID 20239
IUPAC Name 3-{2-[2-(3-azaniumylpropoxy)ethoxy]ethoxy}propan-1-aminium
SMILES [NH3+]CCCOCCOCCOCCC[NH3+]

Specifications

Color Yellow
Boiling Point 151°C
Quantity 100 mL
UN Number 2735
Formula Weight 220.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Bis(3-aminopropyl) Ether
EINECSNumber (7)-1234
RTECSNumber ID6475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.