Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol diethyl ether, 99%

Catalog No. aaa17478ae
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17478AE 100 mL
AAA17478AP 500 mL
AAA174780F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17478AE Supplier Thermo Scientific Chemicals Supplier No. A17478AE
Only null left
Add to Cart

CAS: 112-36-7 | C8H18O3 | 162.229 g/mol

Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.

Solubility
Miscible with hydrocarbons, ethanol, ethyl ether and organic solvents. Immiscible with water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

Melting Point -44°C
Density 0.909
Boiling Point 188°C to 190°C
Flash Point 71°C (160°F)
Odor Weak
Refractive Index 1.412
Linear Formula (CH3CH2OCH2CH2)2O
Quantity 100 mL
Beilstein 1699259
Merck Index 14,3118
Solubility Information Miscible with hydrocarbons,ethanol,ethyl ether and organic solvents. Immiscible with water.
Formula Weight 162.23
Percent Purity 99%
Chemical Name or Material Diethylene glycol diethyl ether
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-963-7
RTECSNumber KN3160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.