Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Dimethacrylate (stabilized with MEHQ) 97.0+%, TCI America™
SDP

Catalog No. D1530100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1530100ML 100 mL
D153025ML 25 mL
D1530500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1530100ML Supplier TCI America Supplier No. D1530100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2358-84-1
Molecular Formula C12H18O5
Molecular Weight (g/mol) 242.27
MDL Number MFCD00014931
InChI Key XFCMNSHQOZQILR-UHFFFAOYSA-N
PubChem CID 16891
IUPAC Name 2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 134°C
Quantity 100 mL
Formula Weight 242.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-1057&(7)-1438
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.